5-Bromo-3-fluorophthalic acid 98%

BLDPBD01445444-100EA 138 EUR
BLDPBD01445444-100 BLDPBD01445444-250 BLDPBD01445444-1G
5-Bromo-3-fluorophthalic acid 98%
5-Bromo-3-fluorophthalic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₄BrFO₄
MW: 263,01 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD32861493
Núm. CAS: 344309-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR