(3aR,8aS)-2-(5-Bromopyridin-2-yl)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole ≥97%, ee 99%

BLDPBD01444816-250EA 160 EUR
BLDPBD01444816-250 BLDPBD01444816-100 BLDPBD01444816-50M BLDPBD01444816-1G
(3aR,8aS)-2-(5-Bromopyridin-2-yl)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole ≥97%, ee 99%
(3aR,8aS)-2-(5-Bromopyridin-2-yl)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₁₅H₁₁BrN₂O
MW: 315,16 g/mol
Temperatura de almacenaje: Nevera
Núm. CAS: 2757083-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR