Nonadecane-1,19-diol 98%

BLDPBD01444523-1GEA 45 EUR
BLDPBD01444523-1G BLDPBD01444523-250 BLDPBD01444523-25G BLDPBD01444523-5G
Nonadecane-1,19-diol 98%
Nonadecane-1,19-diol

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₉H₄₀O₂
MW: 300,5197 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 7268-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR