5,6-Dihydro-1H-cyclobuta[f]inden-4(2H)-one 95+%

BLDPBD01444395-100EA 59 EUR
BLDPBD01444395-100 BLDPBD01444395-250
5,6-Dihydro-1H-cyclobuta[f]inden-4(2H)-one 95+%
5,6-Dihydro-1H-cyclobuta[f]inden-4(2H)-one

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₁H₁₀O
MW: 158,1965 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD24695832
Núm. CAS: 114842-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR