3-Methyl-N-((4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)sulphonyl)butanamide 97%

BLDPBD01444128-1GEA 176 EUR
BLDPBD01444128-1G BLDPBD01444128-5G BLDPBD01444128-100 BLDPBD01444128-250
3-Methyl-N-((4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)sulphonyl)butanamide 97%
3-Methyl-N-((4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)sulphonyl)butanamide

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₇H₂₆BNO₅S
MW: 367,26 g/mol
Temperatura de almacenaje: Nevera
Núm. CAS: 2828439-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR