(3aR,7aR)-1,3-Dimesityl-3a,4,5,6,7,7a-hexahydro-1H-benzo[d]imidazol-3-ium tetrafluoroborate 98%

BLDPBD01442213-1GEA 912 EUR
BLDPBD01442213-1G BLDPBD01442213-250 BLDPBD01442213-100
(3aR,7aR)-1,3-Dimesityl-3a,4,5,6,7,7a-hexahydro-1H-benzo[d]imidazol-3-ium tetrafluoroborate 98%
(3aR,7aR)-1,3-Dimesityl-3a,4,5,6,7,7a-hexahydro-1H-benzo[d]imidazol-3-ium tetrafluoroborate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₂₅H₃₃BF₄N₂
MW: 448,3475 g/mol
Temperatura de almacenaje: Nevera
Núm. CAS: 372517-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR