(SP-4-1)-[[4,4',4'',4'''-(21H,23H-Porphine-5,10,15,20-tetrayl-κN21,κN22,κN23,κN24)tetrakis[benzoato]](6-)]cuprate(4-) 95%

BLDPBD01442000-250EA 144 EUR
BLDPBD01442000-250 BLDPBD01442000-1G BLDPBD01442000-100
(SP-4-1)-[[4,4',4'',4'''-(21H,23H-Porphine-5,10,15,20-tetrayl-κN21,κN22,κN23,κN24)tetrakis[benzoato]](6-)]cuprate(4-) 95%
(SP-4-1)-[[4,4',4'',4'''-(21H,23H-Porphine-5,10,15,20-tetrayl-κN21,κN22,κN23,κN24)tetrakis[benzoato]](6-)]cuprate(4-)

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₄₈H₂₄CuN₄O₈
MW: 848,27 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 64424-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR