1-Cyclopropylpyrrolidin-3-amine dihydrochloride 95%

BLDPBD01440712-1GEA 452 EUR
BLDPBD01440712-1G BLDPBD01440712-100 BLDPBD01440712-250
1-Cyclopropylpyrrolidin-3-amine dihydrochloride 95%
1-Cyclopropylpyrrolidin-3-amine dihydrochloride

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₁₆Cl₂N₂
MW: 199,12 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD28895333
Núm. CAS: 2098058-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR