(R,E)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-phenylhex-5-enoic acid 98%

BLDPBD01440443-1GEA 212 EUR
BLDPBD01440443-1G BLDPBD01440443-250 BLDPBD01440443-100
(R,E)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-phenylhex-5-enoic acid 98%
(R,E)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-phenylhex-5-enoic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₂₇H₂₅NO₄
MW: 427,4917 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD01860907
Núm. CAS: 1272755-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR