Dibenzyl tridecanedioate 95%

BLDPBD01440142-250EA 100 EUR
BLDPBD01440142-250 BLDPBD01440142-1G BLDPBD01440142-100
Dibenzyl tridecanedioate 95%
Dibenzyl tridecanedioate

Storage condition: Sealed in dry, Room Temperature

Formula: C₂₇H₃₆O₄
MW: 424,5724 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 52175-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR