2-Cyclopropyl-5,6,7,8-tetrahydroimidazo[1,2-a]pyridine-8-carboxylic acid hydrochloride 95%

BLDPBD01439550-100EA 94 EUR
BLDPBD01439550-100 BLDPBD01439550-1G BLDPBD01439550-250
2-Cyclopropyl-5,6,7,8-tetrahydroimidazo[1,2-a]pyridine-8-carboxylic acid hydrochloride 95%
2-Cyclopropyl-5,6,7,8-tetrahydroimidazo[1,2-a]pyridine-8-carboxylic acid hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₁H₁₅ClN₂O₂
MW: 242,70 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 2757731-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR