4-((4-(Hydroxymethyl)phenyl)(phenyl)amino)benzaldehyde 95%

BLDPBD01439008-100EA 89 EUR
BLDPBD01439008-100 BLDPBD01439008-1G BLDPBD01439008-250
4-((4-(Hydroxymethyl)phenyl)(phenyl)amino)benzaldehyde 95%
4-((4-(Hydroxymethyl)phenyl)(phenyl)amino)benzaldehyde

Storage condition: Inert atmosphere, Room Temperature

Formula: C₂₀H₁₇NO₂
MW: 303,35 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 334986-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR