2,6-Dichloro-3-methoxyphenol 95%

BLDPBD01438277-250EA 475 EUR
BLDPBD01438277-250 BLDPBD01438277-1G BLDPBD01438277-100 BLDPBD01438277-50M
2,6-Dichloro-3-methoxyphenol 95%
2,6-Dichloro-3-methoxyphenol

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₆Cl₂O₂
MW: 193,0273 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD22376656
Núm. CAS: 86607-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR