(S)-1-(2,3-Dihydro-1H-inden-4-yl)ethan-1-amine hydrochloride 95%

BLDPBD01437597-250EA 463 EUR
BLDPBD01437597-250 BLDPBD01437597-50M BLDPBD01437597-100
(S)-1-(2,3-Dihydro-1H-inden-4-yl)ethan-1-amine hydrochloride 95%
(S)-1-(2,3-Dihydro-1H-inden-4-yl)ethan-1-amine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₁H₁₆ClN
MW: 197,70 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD34186987
Núm. CAS: 2703746-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR