4b1,5,10,15-Tetraazanaphtho[1,2,3-gh]tetraphene-2,3,7,8,12,13-hexaol 95%

BLDPBD01437021-1GEA 1936 EUR
BLDPBD01437021-1G BLDPBD01437021-100 BLDPBD01437021-250
4b1,5,10,15-Tetraazanaphtho[1,2,3-gh]tetraphene-2,3,7,8,12,13-hexaol 95%
4b1,5,10,15-Tetraazanaphtho[1,2,3-gh]tetraphene-2,3,7,8,12,13-hexaol

Storage condition: Inert atmosphere, Room Temperature

Formula: C₂₁H₁₂N₄O₆
MW: 416,34 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 148494-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR