4-Amino-2-(dimethylphosphoryl)benzonitrile 97%

BLDPBD01436940-250EA 37 EUR
BLDPBD01436940-250 BLDPBD01436940-100 BLDPBD01436940-1G
4-Amino-2-(dimethylphosphoryl)benzonitrile 97%
4-Amino-2-(dimethylphosphoryl)benzonitrile

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₉H₁₁N₂OP
MW: 194,1702 g/mol
Temperatura de almacenaje: Nevera
Núm. CAS: 2757731-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR