2-Ethoxy-5-sulphobenzoic acid 98%

BLDPBD01436639-50MEA 271 EUR
BLDPBD01436639-50M BLDPBD01436639-100 BLDPBD01436639-250 BLDPBD01436639-25M BLDPBD01436639-1G
2-Ethoxy-5-sulphobenzoic acid 98%
2-Ethoxy-5-sulphobenzoic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₁₀O₆S
MW: 246,2371 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 2021224-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR