(R)-4-Cyclohexyl-2-(6-cyclopropylpyridin-2-yl)-4,5-dihydrooxazole 97%

BLDPBD01435749-1GEA 796 EUR
BLDPBD01435749-1G BLDPBD01435749-100 BLDPBD01435749-250
(R)-4-Cyclohexyl-2-(6-cyclopropylpyridin-2-yl)-4,5-dihydrooxazole 97%
(R)-4-Cyclohexyl-2-(6-cyclopropylpyridin-2-yl)-4,5-dihydrooxazole

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₇H₂₂N₂O
MW: 270,3694 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 2828438-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR