N,N-Bis(2-ethylhexyl)acrylamide 95%

BLDPBD01435721-100EA 43 EUR
BLDPBD01435721-100 BLDPBD01435721-1G BLDPBD01435721-250
N,N-Bis(2-ethylhexyl)acrylamide 95%
N,N-Bis(2-ethylhexyl)acrylamide

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₉H₃₇NO
MW: 295,5032 g/mol
Temperatura de almacenaje: Nevera
Núm. CAS: 78733-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR