(11bS)-2,6-Bis(3,5-difluorophenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 97%

BLDPBD01435647-250EA 212 EUR
BLDPBD01435647-250 BLDPBD01435647-1G BLDPBD01435647-100
(11bS)-2,6-Bis(3,5-difluorophenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 97%
(11bS)-2,6-Bis(3,5-difluorophenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

Storage condition: Sealed in dry, Room Temperature

Formula: C₃₂H₁₇F₄O₄P
MW: 572,4424 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 1620003-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR