3-((4-Aminophenyl)ethynyl)aniline 95%

BLDPBD01435188-250EA 167 EUR
BLDPBD01435188-250 BLDPBD01435188-100 BLDPBD01435188-1G
3-((4-Aminophenyl)ethynyl)aniline 95%
3-((4-Aminophenyl)ethynyl)aniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₄H₁₂N₂
MW: 208,25 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 85765-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR