Isopropyl 3-bromopropanoate 98%

BLDPBD01434723-100EA 78 EUR
BLDPBD01434723-100 BLDPBD01434723-1G BLDPBD01434723-250
Isopropyl 3-bromopropanoate 98%
Isopropyl 3-bromopropanoate

Storage condition: Sealed in dry, 2-8°C

Formula: C₆H₁₁BrO₂
MW: 195,05 g/mol
Temperatura de almacenaje: Nevera
Núm. CAS: 100983-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR