1-(Cyclopropylmethyl)pyrimidine-2,4(1H,3H)-dione 97%

BLDPBD01433745-100EA 57 EUR
BLDPBD01433745-100 BLDPBD01433745-250 BLDPBD01433745-1G
1-(Cyclopropylmethyl)pyrimidine-2,4(1H,3H)-dione 97%
1-(Cyclopropylmethyl)pyrimidine-2,4(1H,3H)-dione

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₁₀N₂O₂
MW: 166,1772 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD21105338
Núm. CAS: 1462994-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR