(3aS,8aR)-2-(1,10-Phenanthrolin-2-yl)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole 98%

BLDPBD01433437-250EA 303 EUR
BLDPBD01433437-250 BLDPBD01433437-1G BLDPBD01433437-100
(3aS,8aR)-2-(1,10-Phenanthrolin-2-yl)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole 98%
(3aS,8aR)-2-(1,10-Phenanthrolin-2-yl)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole

Storage condition: Inert atmosphere, 2-8°C

Formula: C₂₂H₁₅N₃O
MW: 337,37 g/mol
Temperatura de almacenaje: Nevera
Núm. CAS: 2828438-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR