2-(2,6-Dioxopiperidin-3-yl)-5-iodoisoindoline-1,3-dione 95%

BLDPBD01432559-100EA 73 EUR
BLDPBD01432559-100 BLDPBD01432559-250 BLDPBD01432559-1G
2-(2,6-Dioxopiperidin-3-yl)-5-iodoisoindoline-1,3-dione 95%
2-(2,6-Dioxopiperidin-3-yl)-5-iodoisoindoline-1,3-dione

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₃H₉IN₂O₄
MW: 384,1260 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD31735039
Núm. CAS: 2509137-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR