5,10,15,20-Tetrakis(4-ethylphenyl)porphyrin 95%

BLDPBD01432501-250EA 103 EUR
BLDPBD01432501-250 BLDPBD01432501-1G BLDPBD01432501-100
5,10,15,20-Tetrakis(4-ethylphenyl)porphyrin 95%
5,10,15,20-Tetrakis(4-ethylphenyl)porphyrin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₅₂H₄₆N₄
MW: 726,94 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 146943-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR