(8-Nitro-1,2,3,5,6,7-hexahydro-s-indacen-1-yl)methanol 97%

BLDPBD01432425-100EA 59 EUR
BLDPBD01432425-100
(8-Nitro-1,2,3,5,6,7-hexahydro-s-indacen-1-yl)methanol 97%
(8-Nitro-1,2,3,5,6,7-hexahydro-s-indacen-1-yl)methanol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₃H₁₅NO₃
MW: 233,2631 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 2676864-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR