5-((Trimethylsilyl)ethynyl)thiophene-2-carbaldehyde 98%

BLDPBD01431445-1GEA 166 EUR
BLDPBD01431445-1G BLDPBD01431445-5G BLDPBD01431445-250
5-((Trimethylsilyl)ethynyl)thiophene-2-carbaldehyde 98%
5-((Trimethylsilyl)ethynyl)thiophene-2-carbaldehyde

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₁₀H₁₂OSSi
MW: 208,3522 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD17168296
Núm. CAS: 206768-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR