N4,N4-Bis(4'-amino-[1,1'-biphenyl]-4-yl)-[1,1'-biphenyl]-4,4'-diamine 97%

BLDPBD01431044-100EA 29 EUR
BLDPBD01431044-100 BLDPBD01431044-5G BLDPBD01431044-1G BLDPBD01431044-250
N4,N4-Bis(4'-amino-[1,1'-biphenyl]-4-yl)-[1,1'-biphenyl]-4,4'-diamine 97%
N4,N4-Bis(4'-amino-[1,1'-biphenyl]-4-yl)-[1,1'-biphenyl]-4,4'-diamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₃₆H₃₀N₄
MW: 518,6502 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 2607105-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR