Methyl 5-(dimethylphosphoryl)nicotinate 97%

BLDPBD01430441-250EA 296 EUR
BLDPBD01430441-250 BLDPBD01430441-100 BLDPBD01430441-50M BLDPBD01430441-1G
Methyl 5-(dimethylphosphoryl)nicotinate 97%
Methyl 5-(dimethylphosphoryl)nicotinate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₉H₁₂NO₃P
MW: 213,1702 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD33031725
Núm. CAS: 2613383-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR