1,4,7,10-Tetrakis(pyridin-2-ylmethyl)-1,4,7,10-tetraazacyclododecane 95%

BLDPBD01430164-100EA 30 EUR
BLDPBD01430164-100 BLDPBD01430164-1G BLDPBD01430164-5G BLDPBD01430164-250
1,4,7,10-Tetrakis(pyridin-2-ylmethyl)-1,4,7,10-tetraazacyclododecane 95%
1,4,7,10-Tetrakis(pyridin-2-ylmethyl)-1,4,7,10-tetraazacyclododecane

Storage condition: Inert atmosphere, Room Temperature

Formula: C₃₂H₄₀N₈
MW: 536,7136 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD27965351
Núm. CAS: 185130-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR