5,7-Di-tert-butyl-3-(4-methoxyphenyl)benzo[d]oxazol-3-ium tetrafluoroborate 97%

BLDPBD01430096-1GEA 254 EUR
BLDPBD01430096-1G BLDPBD01430096-100 BLDPBD01430096-250
5,7-Di-tert-butyl-3-(4-methoxyphenyl)benzo[d]oxazol-3-ium tetrafluoroborate 97%
5,7-Di-tert-butyl-3-(4-methoxyphenyl)benzo[d]oxazol-3-ium tetrafluoroborate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₂₂H₂₈BF₄NO₂
MW: 425,26 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 2750161-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR