4-Nitrobenzyl 2-(((tert-butyldimethylsilyl)oxy)methyl)-6,7-dihydropyrazolo[1,5-a]pyrazine-5(4H)-carboxylate 98%

BLDPBD01428684-100EA 180 EUR
BLDPBD01428684-100 BLDPBD01428684-250 BLDPBD01428684-1G
4-Nitrobenzyl 2-(((tert-butyldimethylsilyl)oxy)methyl)-6,7-dihydropyrazolo[1,5-a]pyrazine-5(4H)-carboxylate 98%
4-Nitrobenzyl 2-(((tert-butyldimethylsilyl)oxy)methyl)-6,7-dihydropyrazolo[1,5-a]pyrazine-5(4H)-carboxylate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₂₁H₃₀N₄O₅Si
MW: 446,57 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD34180589
Núm. CAS: 2460029-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR