(R)-4-Benzyl-2-(6-methylpyridin-2-yl)-4,5-dihydrooxazole ≥97%, ee 99%

BLDPBD01428171-250EA 192 EUR
BLDPBD01428171-250 BLDPBD01428171-100 BLDPBD01428171-1G
(R)-4-Benzyl-2-(6-methylpyridin-2-yl)-4,5-dihydrooxazole ≥97%, ee 99%
(R)-4-Benzyl-2-(6-methylpyridin-2-yl)-4,5-dihydrooxazole

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₆H₁₆N₂O
MW: 252,3110 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 2757082-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR