(E)-(E)-Hex-3-en-1-yl hex-3-enoate 95%

BLDPBD01427142-25GEA 92 EUR
BLDPBD01427142-25G BLDPBD01427142-5G BLDPBD01427142-1G
(E)-(E)-Hex-3-en-1-yl hex-3-enoate 95%
(E)-(E)-Hex-3-en-1-yl hex-3-enoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₂₀O₂
MW: 196,2860 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 61444-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR