1H-Indazole-5-carboxylic acid 97%
|
|
BLDPBD01419972-5GEA
32
EUR
BLDPBD01419972-5G
BLDPBD01419972-100
BLDPBD01419972-1G
BLDPBD01419972-25G
1H-Indazole-5-carboxylic acid 97%
1H-Indazole-5-carboxylic acid
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₈H₆N₂O₂ MW: 162,15 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD06804570 Núm. CAS: 61700-61-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...