2-Methyl-2H-indazole-5-boronic acid pinacol ester 98%

BLDPBD01406911-5GEA 572 EUR
BLDPBD01406911-5G BLDPBD01406911-250 BLDPBD01406911-100 BLDPBD01406911-1G
2-Methyl-2H-indazole-5-boronic acid pinacol ester 98%
2-Methyl-2H-indazole-5-boronic acid pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₄H₁₉BN₂O₂
MW: 258,1239 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD11109436
Núm. CAS: 1189746-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR