2,4-Dichloro-L-phenylalanine 98+%
|
|
BLDPBD01397815-1GEA
19
EUR
BLDPBD01397815-1G
BLDPBD01397815-5G
BLDPBD01397815-100
BLDPBD01397815-25G
2,4-Dichloro-L-phenylalanine 98+%
2,4-Dichloro-L-phenylalanine
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₉H₉Cl₂NO₂ MW: 234,1 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD01860882 Núm. CAS: 111119-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...