DL-Methionine sulphoxide 98+%

Proveedor: BLD PHARMATECH GMBH

BLDPBD01393110-5GEA 82 EUR
BLDPBD01393110-5G BLDPBD01393110-1G BLDPBD01393110-100 BLDPBD01393110-25G
DL-Methionine sulphoxide 98+%
DL-Methionine sulphoxide

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₅H₁₁NO₃S
MW: 165,21 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00002620
Núm. CAS: 62697-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR