Iminodiacetonitrile 92%

BLDPBD01384053-25GEA 7 EUR
BLDPBD01384053-25G
Iminodiacetonitrile 92%
Iminodiacetonitrile

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₄H₅N₃
MW: 95,11 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00001887
Núm. CAS: 628-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR