2,3-Difluoro-4-ethoxybenzonitrile 98+%

BLDPBD01382258-5GEA 37 EUR
BLDPBD01382258-5G BLDPBD01382258-100 BLDPBD01382258-1G BLDPBD01382258-25G
2,3-Difluoro-4-ethoxybenzonitrile 98+%
2,3-Difluoro-4-ethoxybenzonitrile

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₇F₂NO
MW: 183,1548 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD04973770
Núm. CAS: 126162-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR