4-Bromo-N-(3-bromophenyl)benzenesulphonamide 95%

BLDPBD00952046-1GEA 185 EUR
BLDPBD00952046-1G BLDPBD00952046-100 BLDPBD00952046-250
4-Bromo-N-(3-bromophenyl)benzenesulphonamide 95%
4-Bromo-N-(3-bromophenyl)benzenesulphonamide

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₉Br₂NO₂S
MW: 391,0784 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD01215114
Núm. CAS: 349404-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR