Nitazoxanide 97%

Sinónimos: NTZ

BLDPBD00938966-500EA 227 EUR
BLDPBD00938966-500 BLDPBD00938966-25G BLDPBD00938966-10G BLDPBD00938966-0.5 BLDPBD00938966-100 BLDPBD009389661000
Nitazoxanide 97%
Nitazoxanide

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₂H₉N₃O₅S
MW: 307,28 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00416599
Núm. CAS: 55981-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR