5-(4-Hydroxyphenyl)-1H-tetrazole 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD00807563-5GEA 335 EUR
BLDPBD00807563-5G BLDPBD00807563-250 BLDPBD00807563-1G
5-(4-Hydroxyphenyl)-1H-tetrazole 98%
5-(4-Hydroxyphenyl)-1H-tetrazole

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₆N₄O
MW: 162,16 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD05662709
Núm. CAS: 51517-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR