Erucic acid 85%

BLDPBD00806303-1GEA 0 EUR
BLDPBD00806303-1G BLDPBD00806303100G BLDPBD00806303-25G BLDPBD008063031000 BLDPBD00806303-5G BLDPBD00806303-500 BLDPBD00806303-10G
Erucic acid 85%
Erucic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₂₂H₄₂O₂
MW: 338,58 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00063188
Núm. CAS: 112-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR