3-Fluoro-2-methoxy-5-pyridineboronic acid 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD00771619-250EA 45 EUR
BLDPBD00771619-250 BLDPBD00771619-100 BLDPBD00771619-1G BLDPBD00771619-5G
3-Fluoro-2-methoxy-5-pyridineboronic acid 98%
3-Fluoro-2-methoxy-5-pyridineboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₆H₇BFNO₃
MW: 170,9341 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD04112520
Núm. CAS: 856250-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR