8-(Bromomethyl)isoquinoline hydrobromide 97%

BLDPBD00743653-250EA 173 EUR
BLDPBD00743653-250 BLDPBD00743653-100
8-(Bromomethyl)isoquinoline hydrobromide 97%
8-(Bromomethyl)isoquinoline hydrobromide

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₀H₈BrN·BrH
MW: 302,993 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD12674806
Núm. CAS: 1215541-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR