1,4-Dicyano-2-butene 99+%

Proveedor: BLD PHARMATECH GMBH

BLDPBD00727789-1GEA 7 EUR
BLDPBD00727789-1G BLDPBD00727789-25G BLDPBD00727789-5G
1,4-Dicyano-2-butene 99+%
1,4-Dicyano-2-butene

Storage condition: Sealed in dry, Room Temperature

Formula: C₆H₆N₂
MW: 106,1252 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00001964
Núm. CAS: 1119-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR