Powered air respirator filter cover for Versaflo™ TR-6310E, Versaflo™
Proveedor: 3M
Versaflo™
MMMATR6300FCEA 27.2 EUR
MMMATR6300FC
Powered air respirator filter cover for Versaflo™ TR-6310E, Versaflo™
Respiradores Sistemas de respiración
The Versaflo™ Filter Cover TR-6300FC is a filter cover which helps protect 3M™ Versaflo™ TR-6310E filter from physical damage.
  • Protects filter from physical damage
  • Can be cleaned and reused
This filter cover safeguards the filter and enables users to wear Versaflo™ Powered Air Turbo through decontamination showers. It can hold a Versaflo™ Spark Arrestor TR-662, or Versaflo™ Prefilter TR-6600 in place for additional protection from larger particles or sparks. The filter cover can be cleaned for reuse.