Parker Hannifin Threaded Reducing Nipples, NPT(M) x NPT(M)
Proveedor: Parker Hannifin
MFLX31321-07EA 34.5 EUR
MFLX31321-07 MFLX31321-06 MFLX31321-05 MFLX31321-04 MFLX31321-03 MFLX31321-02 MFLX31321-01 MFLX31320-98 MFLX31321-00 MFLX31320-99
Parker Hannifin Threaded Reducing Nipples, NPT(M) x NPT(M)
Racores y conectores
Create a secure connection with threaded fittings.
  • Constructed of chemically inert PFA
Threaded fittings and connections offer a more permanent installation. Use these nipples to reduce or enlarge connection sizes.